Learn More
Silica gel desiccant, indicating, ACS, Thermo Scientific Chemicals
Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits.
$75.55 - $1136.90
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4038122
|
Thermo Scientific Chemicals
04038122 |
100 g |
Each for $75.55
|
|
|||||
AA4038136
|
Thermo Scientific Chemicals
04038136 |
500 g |
Each for $255.67
|
|
|||||
AA40381A4
|
Thermo Scientific Chemicals
040381A4 |
2.5 kg |
Each for $1,136.90
|
|
|||||
Description
Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
24261 | |
dioxosilane |
Specifications
Granular | |
7631-86-9 | |
Silica Gel | |
O2Si | |
Odorless | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
Hygroscopic | |
60.08 | |
CHEBI:30563 | |
Silica gel desiccant, For analysis ACS |
Blue | |
2.17 to 2.66g/cm3 | |
100 g | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Soluble in alkalies. Insoluble in water. | |
O=[Si]=O | |
dioxosilane | |
24261 | |
ACS |
Safety and Handling
P201-P202-P281-P308+P313-P501c
H350i-H360F
EINECSNumber : 231-545-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only