missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silica gel, for column chrom., ultrapure, 60-200μ, 60A, Thermo Scientific Chemicals
$123.19 - $3195.44
Chemical Identifiers
CAS | 7631-86-9 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | Silicon dioxide |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC360062500
|
Thermo Scientific Chemicals
360062500 |
250 g | Plastic Bottle |
Each for $123.19
|
|
||||
AC360060010
|
Thermo Scientific Chemicals
360060010 |
1 kg | Plastic Bottle |
Each for $409.42
|
|
||||
AC360060050
|
Thermo Scientific Chemicals
360060050 |
5 kg | Plastic Drum |
Each for $1,504.92
|
|
||||
AC360060300
|
Thermo Scientific Chemicals
360060300 |
30 kg | Plastic Bottle |
Each for $3,195.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
7631-86-9 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
O=[Si]=O |
O2Si | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
Silicon dioxide | |
CHEBI:30563 |
Specifications
Powder and/or Chunks | |
6.5 to 7.5 (10% w/w suspension) | |
Silica Gel | |
O2Si | |
Plastic Bottle | |
Silicon dioxide | |
Solubility in water: insoluble. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Silica gel, For column chromatography |
White to Brown | |
7631-86-9 | |
250 g | |
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 | |
5.0% Max. (under size) 5.0% Max. (over size) | |
0.70 to 0.85mL/g (specific) | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 |
RUO – Research Use Only