MilliporeSigma™ Silica gel 60 GF254 for thin-layer chromatography, MilliporeSigma™
Silica gel 60 GF254 for thin-layer chromatography. Silica gel 60 GF254 CAS No. 7631-86-9, EC Number 231-545-4.
$148.59 - $4277.49
Specifications
Grade 60
Type Packing Material
Material Silica Gel
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1077301000
View Documents Promotion Details
MilliporeSigma™
1077301000
1 kg Plastic Bottle
Each for $148.59
M1077305000
View Documents Promotion Details
MilliporeSigma™
1077305000
5 kg Plastic Drum
Each for $617.48
M1077309025
View Documents Promotion Details
MilliporeSigma™
1077309025
25 kg Plastic Drum
Each for $4,277.49
Description
Description
Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40μm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).
Specifications
Specifications
60
Packing Material
Silica Gel
SDS
missing translation for 'documents'
missing translation for 'documents'