missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps

Optimize cell yield with the Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps.

Manufacturer:  Thermo Scientific™ A78710020

 View more versions of this product

Catalog No. A78710020



Description

Description

Optimize cell yield with the Thermo Scientific™ Shandon™ EZ Single Cytofunnel™ with White Filter Cards and Caps. A disposable sample chamber with cap and pre-attached white filter card for high absorbency, this product is meant for single use with all Thermo Scientific™ Shandon™ Cytospin™ Cytocentrifuge applications.

  • Reduces the risk of exposure to pathogenic samples
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Decreases the possibility of cross-contamination
  • Eliminates the need for metal Thermo Scientific™ Shandon™ Cytoclips™
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Holds sample volumes up to 0.5mL
  • Includes cell deposition area of 6mm in diameter
Includes:
  • Closure caps
  • Easy-to-use operational illustrations
Specifications

Specifications

200/Cs.
200/Cs
Disposable|With Filter Cards and Caps
White
Documents