Thermo Scientific™ Shandon™ Double Cytofunnel™
Optimize cell yield with the Thermo Scientific™ Shandon™ Double Cytofunnel™.
Manufacturer:  Thermo Scientific™ 5991039
 View more versions of this product
Catalog No. 59-910-39
Description
Description
Optimize cell yield with the Thermo Scientific™ Shandon™ Double Cytofunnel™. Features disposable sample chambers with white filter cards and caps.
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Reduces the risk of exposure to pathogenic samples
  • Reduces preparation time
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Eliminates the need for metal Thermo Scientific™ Shandon™ Cytoclips™
  • Decreases the possibility of cross-contamination
  • Features funnel markings corresponding to slide markings for simple identification
Includes: 
  • Pre-attached filter card
  • Closure caps
  • Easy-to-use operational illustrations
Specifications
Specifications
25/Box
25/Box
Disposable Sample Chamber|With Filter Cards and Caps
Documents