SGE™ Series II Standard Fixed Needle Syringes
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
$38.17 - $212.19
Specifications
Accuracy ±1%
Diameter (English) Outer Needle 0.01 in.
Length (English) Needle 1.96 in.
Length (Metric) Needle 50mm
Material (Barrel) Borosilicate Glass
View More Specs
Products 8
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Includes Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Includes Price Quantity  
SG-001000 View Documents SGE™
SG001000
5μL 26 ga. Plunger protection Each for $69.24
SG-002000 View Documents SGE™
SG002000
10μL 26 ga. Plunger protection Each for $38.17
SG-002030 View Documents SGE™
SG002030
10μL 26 ga. Plunger protection Pack of 6 Each for $212.19
SG-003000 View Documents SGE™
SG003000
25μL 25 ga. Standard plunger Each for $56.74
SG-004000 View Documents SGE™
SG004000
50μL 25 ga. Standard plunger Each for $77.15
SG-005000 View Documents SGE™
SG005000
100μL 25 ga. Standard plunger Each for $71.56
SG-006000 View Documents SGE™
SG006000
250μL 25 ga. Standard plunger Each for $88.65
SG-007000 View Documents SGE™
SG007000
500μL 25 ga. Standard plunger Each for $95.72
Description
Description
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Specifications
±1%
1.96 in.
Borosilicate Glass
0.01 in.
50mm