SGE™ BPX50 Capillary Column
Offers a comprehensive GC capillary column range for almost any chromatographic application
Manufacturer:  SGE™ SG054740
 View more versions of this product
Catalog No. SG054740
Description
Description
Use for EPA methods 604, 608, 8060, 8081, triazines/herbicides, drug screening, steroids and a variety of pharmaceutical applications
  • Mid polarity
  • Inert
  • Low bleed
  • High temperature
Documents