missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads
Suited for capturing or isolating mRNA from a variety of sources. Due to their “cauliflower like” structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.
$244.37 - $5307.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | mRNA Purification |
Diameter (Metric) | 1 μm |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
09-880-012
|
Cytiva
38152103011150 |
1 mL |
Each for $244.37
|
|
|||||
09981145
|
Cytiva
38152103010150 |
5 mL |
Each for $942.00
|
|
|||||
09-880-014
|
Cytiva
38152103010350 |
100 mL |
Each for $5,307.00
|
|
|||||
Description
- Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
- High binding capacity
- 1.0μm diameter
- High oligo dT content for mRNA binding
Specifications
1% solids (10mg/mL) | |
2°C to 8°C | |
mRNA Purification | |
1 μm |