missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Selenium, plasma standard solution, Specpure™ Se 1000μg/mL, Thermo Scientific Chemicals
Se 1000μg/mL
$85.45 - $120.92
Chemical Identifiers
CAS | 7697-37-2 |
---|---|
Molecular Formula | HNO3 |
Molecular Weight (g/mol) | 63.01 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
PubChem CID | 944 |
ChEBI | CHEBI:48107 |
IUPAC Name | nitric acid |
SMILES | O[N+]([O-])=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7697-37-2 | |
63.01 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
CHEBI:48107 | |
O[N+]([O-])=O |
HNO3 | |
MFCD00011349 | |
944 | |
nitric acid |
Specifications
7697-37-2 | |
HNO3 | |
GRYLNZFGIOXLOG-UHFFFAOYSA-N | |
nitric acid | |
944 | |
Nitric acid: 5%, Selenium: 0.15%, Water: 94.9% | |
Liquid |
50 mL | |
MFCD00011349 | |
O[N+]([O-])=O | |
63.01 | |
CHEBI:48107 | |
23 hPa (17mm Hg) at 20°C | |
Selenium, plasma standard solution |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: III
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only