missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Science Education™ Seamless Cellulose Dialysis Tubing
Retains materials with 12,000 and higher molecular weight.
$28.26 - $264.41
Specifications
Product Type | Dialysis Tubing |
---|---|
Material | Seamless Cellulose |
Formulation | Glycerin, water and 1% sulfur |
Quantity | Each |
For Use With (Application) | Useful in analytical chemistry, biology, and pharmaceutical fields |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (English) | Length (Metric) | Price | Quantity | |||
S25645F
|
Fisher Science Education™
S25645F |
15.9 mm | 100 ft. | 30 m |
Each for $234.95
|
|
|||
S25645B
|
Fisher Science Education™
S25645B |
28.6 mm | 10 ft. | 3 m |
Each for $33.69
|
|
|||
S25645A
|
Fisher Science Education™
S25645A |
20.4 mm | 10 ft. | 3 m |
Each for $31.08
|
|
|||
S25645
|
Fisher Science Education™
S25645 |
15.9 mm | 10 ft. | 3 m |
Each for $28.26
|
|
|||
S25645C
|
Fisher Science Education™
S25645C |
47.7 mm | 10 ft. | 3 m |
Each for $87.49
|
|
|||
S25645D
|
Fisher Science Education™
S25645D |
6.4 mm | 10 ft. | 3 m |
Each for $78.64
|
|
|||
S25645G
|
Fisher Science Education™
S25645G |
32 mm | 100 ft. | 30 m |
Each for $264.41
|
|
|||
Description
- Especially useful in analytical chemistry, biology, and pharmaceutical fields
- Highly permeable to water and low molecular weight compounds
- Contains glycerin, water and 1% sulfur
- Must be stored in airtight container or rehumidified before use
- Pore diameter: 4.8nm
This product is for educational use - no C of A is available.
Specifications
Dialysis Tubing | |
Glycerin, water and 1% sulfur | |
Useful in analytical chemistry, biology, and pharmaceutical fields |
Seamless Cellulose | |
Each |