missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Scandium, plasma standard solution, Specpure™ Sc 1000μg/mL, Thermo Scientific Chemicals
Sc 1000μg/mL
$225.03 - $225.03
Chemical Identifiers
CAS | 7440-20-2 |
---|---|
Molecular Formula | Sc |
Molecular Weight (g/mol) | 44.96 |
MDL Number | MFCD00016323 |
InChI Key | SIXSYDAISGFNSX-UHFFFAOYSA-N |
PubChem CID | 23952 |
ChEBI | CHEBI:33330 |
IUPAC Name | scandium |
SMILES | [Sc] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA35769AE
|
Thermo Scientific Chemicals
035769AE |
100 mL |
Each for $225.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-20-2 | |
44.96 | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
CHEBI:33330 | |
[Sc] |
Sc | |
MFCD00016323 | |
23952 | |
scandium |
Specifications
7440-20-2 | |
Sc | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
scandium | |
23952 | |
Nitric acid: 5%, Scandium(III) oxide: 0.15%, Water: 94.85% | |
Liquid |
100 mL | |
MFCD00016323 | |
[Sc] | |
44.96 | |
CHEBI:33330 | |
23 hPa (17mm Hg) at 20°C | |
Scandium, plasma standard solution |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only