missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Savant™ SPD121P SpeedVac™ Kits

Conduct combinatorial chemistry applications Thermo Scientific™ SPD121P SpeedVac™ kits, which resist TFA, DSMO and other solvents.

Manufacturer:  Thermo Scientific™ SPD121PP1-115

 View more versions of this product

Catalog No. 11-716-806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Conduct combinatorial chemistry applications Thermo Scientific™ SPD121P SpeedVac™ kits, which resist TFA, DSMO and other solvents. All of our SpeedVac kits contain all the necessary components under one model number.

Features:

  • Deep aluminum standard PTFE-coated chamber
  • Safety cover lock
  • Radiant glass safety cover improves drying time by applying radiant IR heat to samples
  • PTFE plumbing and tubing
  • Two valve manifold with built-in automatic bleeder valve
  • Time/temperature display incorporating membrane touch pad switches
  • Variable drying times from 35° to 80°C in five degree increments
  • Manual/auto timed run
  • Endpoint mode terminates run when pre-set vacuum gauge and vacuum displays

Solvents and Solvent Combinations:

  • Acetone
  • Chloroform
  • Ethyl acetate
  • Hexane
  • Methylene chloride

Includes:

  • SpeedVac Concentrator, Enhanced Evaporation
  • -104°C, Ultra-Low Temperature Refrigerated Trap
  • OFP400 Oil-Free ETFE/PFA 4-Headed Pump
  • 64 × 1.5mL Rotor
  • Digital Vacuum Gauge
  • Chemical Trap Kit
  • Disposable Cartridge
  • Tubing and Fitting Kit
  • Glass Condensation Flask
  • CryoCool Heat Transfer Fluid (1L)

Recommended for:

  • Removing ethyl acetate from samples for CAT assay
  • Drug metabolities in solid phase extracts
  • Analyte elution solvents for evaporation
Specifications

Specifications

SPD121P-115
PTFE-coated Aluminum
115V
34 in.
82 in.
32 in.
355 lb.
Savant SpeedVac Concentrator Kits, Enhanced Evaporation; 115V/60Hz
115V 60Hz
35°C to 80°C in 5 degree increments
0.1 to 20 torr endpoint shut off
60Hz
864mm
2083mm
813mm
164kg
82 x 32 x 34 in. (2083 x 813 x 864mm)
Documents