missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Supplier:  Sartorius VS0272

 View more versions of this product

Catalog No. 14-558-432



Description

Description

Combines high speed with superior retentate recovery for concentrating very dilute solutions. Sartorius Vivaspin™ 2 Centrifugal Concentrators: PES Membrane feature twin vertical membranes, small internal surface areas and thin-channel chambers. Available in a range of MWCO and packaging quantities.

Vivaspin concentrators are disposable ultrafiltration devices for concentrating biological samples. Vivaspin 2 is designed for use with samples from 500μl to 2mL in rotors that accept 15mL centrifuge tubes. The unique design provides high-speed concentrations for even particle-laden solutions and near-total concentrate recovery.

Twin vertical membranes and optimal design for fast, safe concentration
  • Patented twin vertical membrane design for ultra-fast processing speeds
  • In a single spin, solution volumes up to 4mL can be concentrated up to 30x in 5–30 minutes
  • Thin-channel filtration chamber minimizes membrane fouling and increases filtration speed
  • Dead stop pocket built into the bottom eliminates the risk of sample filtration to dryness
  • High-flux PES membranes feature very low protein binding and low extractables
Designed for maximum flexibility and sample recovery
  • Low internal surface and membrane area for superior recoveries from very dilute solutions
  • Two methods for easy concentrate recovery:
    • Directly pipet the concentrate from the dead stop pocket in the concentrator
    • Reverse-spin the concentrate into the recovery cap, which can be sealed for storage
    • Both methods provide near-total concentrate recovery of greater than 90%
  • Volume graduation for easy monitoring of concentrate volume and 8μl dead-stop volume
  • Universal compatibility with swing bucket or fixed angle rotors accepting 15mL centrifuge tubes
Specifications

Specifications

Single-use
0.2 μm
Concentration, desalting and buffer applications
Swing bucket rotor: 4,000 x g
Fixed angle rotor: 9,000 x g (3 - 50kDa MWCO); 12,000 x g (100kDa+ MWCO)
2 mL (Fixed Angle Rotor), 3 mL (Swing Bucket Rotor)
17.8 mm
PES
1.3 cm2
Polycarbonate
100/Pk.
126 mm
SDS
Documents

Documents

Promotions

Promotions