Sartorius™ UniVessel™ SU Accessories: Motor Adaptor
Accessories for Single-Use UniVessel™ SU Stirred Tank Bioreactor
$345.51 - $345.51
Specifications
Disposable Yes
For Use With Univessel™ su
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
14-559-232 View Documents Sartorius™
DZ-------MAP
Motor adaptor - Aplus Each for $345.51
14-559-231 View Documents Sartorius™
DZMB
Motor adaptor - Bplus / B-DCU I / B-DCU II Each for $345.51
14-559-233 View Documents Sartorius™
DZ-------MAA
Motor adaptor - Applikon Each for $345.51
Description
Description
  • Used with most bioreactor controllers for autoclavable bioreactors
  • Stainless-steel adaptors for several existing motors are available
  • Mounted on UniVessel SU stirrer shaft coupling
  • Features bayonet lock for secure motor-vessel connection
  • Compatible with Biostat Aplus, Biostat Bplus, new Biostat B and Biostat B DCU-I and DCU-II
Specifications
Specifications
Yes
Univessel™ su