Learn More
Sartorius Cellulose Nitrate Membrane Filters
Combines uniform pore structure with high flow and adsorption rates. Sartorius Cellulose Nitrate Membrane Filters are excellently suited for diagnostic kits. Non-sterile, hydrophilic filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
$131.97 - $780.47
Specifications
Autoclavable | Autoclavable (wet) |
---|---|
Color | White |
Material | Cellulose Nitrate |
Hydrophobicity | Hydrophilic |
Thickness (Metric) | 130 μm |
Alert:
Cellulose nitrate filters are highly flammable. Their flashpoint is approx. 200°C. Always keep these filters separated by the discs of interleaving paper that are supplied with the filters.
Catalog Number | Mfr. No. | Diameter (Metric) | Pore Size | Flow Rate | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Pore Size | Flow Rate | Price | Quantity | |||
14-555-375
|
Sartorius
1130125N |
25 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-372
|
Sartorius
11301100G |
100 mm | 8 μm | 750mL/min. |
Pack of 25 Each for $274.25
|
|
|||
14-555-373
|
Sartorius
11301100N |
100 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $780.47
|
|
|||
14-555-374
|
Sartorius
1130113N |
13 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $131.97
|
|
|||
14-555-376
|
Sartorius
1130137N |
37 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $224.76
|
|
|||
14-555-377
|
Sartorius
1130147N |
47 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $285.59
|
|
|||
14-555-380
|
Sartorius
1130213N |
13 mm | 3 μm | 430mL/min. |
Pack of 100 Each for $131.97
|
|
|||
14-555-383
|
Sartorius
1130250N |
50 mm | 3 μm | 430mL/min. |
Pack of 100 Each for $246.41
|
|
|||
14-555-379
|
Sartorius
11302100G |
100 mm | 3 μm | 430mL/min. |
Pack of 25 Each for $276.31
|
|
|||
14-555-384
|
Sartorius
11303100G |
100 mm | 1.2 μm | 200mL/min. |
Pack of 25 Each for $246.41
|
|
|||
14-555-386
|
Sartorius
1130347N |
47 mm | 1.2 μm | 200mL/min. |
Pack of 100 Each for $285.59
|
|
|||
14-555-378
|
Sartorius
1130150N |
50 mm | 8 μm | 750mL/min. |
Pack of 100 Each for $246.41
|
|
|||
14-555-385
|
Sartorius
1130325N |
25 mm | 1.2 μm | 200mL/min. |
Pack of 100 Each for $156.71
|
|
|||
14-555-387
|
Sartorius
1130350N |
50 mm | 1.2 μm | 200mL/min. |
Pack of 100 Each for $246.41
|
|
|||
14-555-388
|
Sartorius
11304100G |
100 mm | 0.8 μm | 200mL/min. |
Pack of 25 Each for $249.51
|
|
|||
14-555-389
|
Sartorius
1130413N |
13 mm | 0.8 μm | 200mL/min. |
Pack of 100 Each for $131.97
|
|
|||
14-555-392
|
Sartorius
1130447N |
47 mm | 0.8 μm | 200mL/min. |
Pack of 100 Each for $249.51
|
|
|||
14-555-397
|
Sartorius
1130550N |
50 mm | 0.65 μm | 130mL/min. |
Pack of 100 Each for $204.14
|
|
|||
14-555-396
|
Sartorius
1130547N |
47 mm | 0.65 μm | 130mL/min. |
Pack of 100 Each for $251.58
|
|
|||
14-555-390
|
Sartorius
1130425N |
25 mm | 0.8 μm | 200mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-395
|
Sartorius
1130525N |
25 mm | 0.65 μm | 130mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-398
|
Sartorius
11306100G |
100 mm | 0.45 μm | 70mL/min. |
Pack of 25 Each for $296.93
|
|
|||
14-555-399
|
Sartorius
1130613N |
13 mm | 0.45 μm | 70mL/min. |
Pack of 100 Each for $156.71
|
|
|||
14-555-404
|
Sartorius
1130690N |
90 mm | 0.45 μm | 70mL/min. |
Pack of 100 Each for $780.47
|
|
|||
14-555-400
|
Sartorius
1130625N |
25 mm | 0.45 μm | 70mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-402
|
Sartorius
1130650N |
50 mm | 0.45 μm | 70mL/min. |
Pack of 100 Each for $204.14
|
|
|||
14-555-405
|
Sartorius
1134213N |
13 mm | 5 μm | 570mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-401
|
Sartorius
1130647N |
47 mm | 0.45 μm | 70mL/min. |
Pack of 100 Each for $204.14
|
|
|||
14-555-403
|
Sartorius
1130690G |
90 mm | 0.45 μm | 70mL/min. |
Pack of 25 Each for $204.14
|
|
|||
14-555-406
|
Sartorius
1134225N |
25 mm | 5 μm | 570mL/min. |
Pack of 100 Each for $224.76
|
|
|||
14-555-407
|
Sartorius
1134247N |
47 mm | 5 μm | 570mL/min. |
Pack of 100 Each for $296.93
|
|
|||
14-555-408
|
Sartorius
1134250N |
50 mm | 5 μm | 570mL/min. |
Pack of 100 Each for $308.27
|
|
|||
14-555-393
|
Sartorius
1130450N |
50 mm | 0.8 μm | 200mL/min. |
Pack of 100 Each for $249.51
|
|
|||
14-555-381
|
Sartorius
1130225N |
25 mm | 3 μm | 430mL/min. |
Pack of 100 Each for $175.27
|
|
|||
14-555-382
|
Sartorius
1130247N |
47 mm | 3 μm | 430mL/min. |
Pack of 100 Each for $224.76
|
|
|||
Description
Combines uniform pore structure with high flow and adsorption rates. Sartorius Cellulose Nitrate Membrane Filters are excellently suited for diagnostic kits. Non-sterile, hydrophilic filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
- Cellulose nitrate filters combine high flow rates with high non-specific adsorption
- Defined particle retention and outstanding, reliable capture rates for microorganisms
- Very advantageous for diagnostic kits and compatible with aqueous solutions, hydrocarbons and several other organic solvents
- Filters have a uniform pore structure and are available in range of diameters and pore sizes
- Pore sizes: 0.45μm, 0.65μm, 0.8μm, 1.2μm, 3.0μm, 5μm and 8μm
- The larger pore sizes (8 μm, 5 μm, 3 μm) are useful for cell retention and prefiltering
- The 0.45 pore size is ideal for particle collection and filtration
- Diameters: 13, 25, 37, 47, 50, 90 and 100mm (0.5 to 3.9 in.)
- Ungridded filters can be sterilized via autoclave (wet) to 121°C, ethylene oxide or γ-irradiation to 25 kGy
- Box label specifies the type, quantity, diameter, pore size and lot number
Specifications
Autoclavable (wet) | |
Cellulose Nitrate | |
130 μm |
White | |
Hydrophilic |