missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salinity Refractometer

Portable device measures the percentage of salt in any body of water.

Manufacturer:  Fisher Scientific 78620

Catalog No. S94223



Includes: Two pipets, calibration screwdriver, cotton cleaning cloth, vial cleaning solution, hard plastic case and instructions

Description

Description

  • Solid aluminum construction with rubber-gripped body
  • Automatic temperature compensation
  • Prism and lens feature simple focus adjustments
  • Accurate results with only two to three drops of solution
  • Dimensions: 1.5D × 7 in.L (3.8 × 17.7cm)
  • Range: 0-28%
  • Resolution: 0.2%
Specifications

Specifications

Two pipets, calibration screwdriver, cotton cleaning cloth, vial cleaning solution, hard Plastic case and instructions
0.2%
0-28%
7 in.
177.8mm
Documents