missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ SafTest™ Analyzer Liquid Sample Instrument Platform

Used in conjunction with alkenal, peroxide test kit.

Specifications

Storage Requirements Room temperature
View More Specs

Includes: Timer, two block dri-bath, insert, vortex, test tube rack, pipette PDP, vacuum pump assembly, acrylic base, 100 to 1000μL pipette PDP, 15mm dri-block insert, 20mm test tube rack, SafTest analyzer, tube rocker, O-ring for vacuum pump assembly

Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
MP07LI2000 View Documents MP Biomedicals™
07LI2000
Used for glycerin, malonaldehyde testing N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MP07LI2020 View Documents MP Biomedicals™
07LI2020
Used for peroxide, alkenal testing N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Equipment platform for conducting tests
  • Contains all of instrumentation necessary to run solid samples
  • Test time: 30 minutes
  • Research area: Lipid degradation and oxidation
  • Detection method: Bichromatic photometer
  • Dimensions: 30 x 27.2 x 12.5cm
Specifications

Specifications

Room temperature