missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(S)-(-)-Azetidine-2-carboxylic acid, 99%, Thermo Scientific Chemicals
$240.23 - $610.14
Chemical Identifiers
CAS | 2133-34-8 |
---|---|
Molecular Formula | C4H7NO2 |
Molecular Weight (g/mol) | 101.105 |
MDL Number | MFCD00005166 |
InChI Key | IADUEWIQBXOCDZ-VKHMYHEASA-N |
Synonym | l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l |
PubChem CID | 16486 |
ChEBI | CHEBI:6198 |
IUPAC Name | (2S)-azetidine-2-carboxylic acid |
SMILES | C1CNC1C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL09668MD
|
Thermo Scientific Chemicals
L09668MD |
250 mg |
Each for $240.23
|
|
|||||
AAL0966803
|
Thermo Scientific Chemicals
L0966803 |
1 g |
Each for $610.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2133-34-8 | |
101.105 | |
IADUEWIQBXOCDZ-VKHMYHEASA-N | |
16486 | |
(2S)-azetidine-2-carboxylic acid |
C4H7NO2 | |
MFCD00005166 | |
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l | |
CHEBI:6198 | |
C1CNC1C(=O)O |
Specifications
2133-34-8 | |
C4H7NO2 | |
250 mg | |
14,908 | |
IADUEWIQBXOCDZ-VKHMYHEASA-N | |
C1CNC1C(=O)O | |
101.105 | |
CHEBI:6198 | |
99% |
∽217°C (decomposition) | |
MFCD00005166 | |
80678 | |
l-azetidine-2-carboxylic acid, s-azetidine-2-carboxylic acid, s---2-azetidinecarboxylic acid, 2s-azetidine-2-carboxylic acid, azetidyl-2-carboxylic acid, l-azetidine 2-carboxylic acid, s-2-azetidinecarboxylic acid, 2-azetidinecarboxylic acid, s, s---azetidine-2-carboxylic acid, 2-azetidinecarboxylic acid, l | |
−123° (c=4 in H2O) | |
(2S)-azetidine-2-carboxylic acid | |
16486 | |
101.11 | |
(S)-(-)-Azetidine-2-carboxylic acid |
Safety and Handling
EINECSNumber : 218-362-5
RTECSNumber : CM4310500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only