missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rutin Hydrate, 97+%, Thermo Scientific Chemicals
$76.00 - $755.18
Chemical Identifiers
CAS | 207671-50-9 |
---|---|
Molecular Formula | C27H30O16 |
Molecular Weight (g/mol) | 610.52 |
MDL Number | MFCD01319140 |
InChI Key | IKGXIBQEEMLURG-NVPNHPEKSA-N |
Synonym | rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid |
PubChem CID | 5280805 |
ChEBI | CHEBI:28527 |
IUPAC Name | 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
SMILES | C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC132390050
|
Thermo Scientific Chemicals
132390050 |
5 g | Glass Bottle |
Each for $76.00
|
|
||||
AC132391000
|
Thermo Scientific Chemicals
132391000 |
100 g | Plastic bottle |
Each for $242.41
|
|
||||
AC132395000
|
Thermo Scientific Chemicals
132395000 |
500 g | Plastic bottle |
Each for $755.18
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207671-50-9 | |
610.52 | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
5280805 | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
C27H30O16 | |
MFCD01319140 | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
CHEBI:28527 | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O |
Specifications
207671-50-9 | |
White | |
Authentic | |
Glass Bottle | |
MFCD01319140 | |
15, 8438 | |
Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O | |
610.52 | |
CHEBI:28527 | |
97+% | |
Rutin hydrate, 97.0-102.0% |
195.0°C | |
5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum) | |
97+% | |
C27H30O16 | |
5 g | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
5280805 | |
664.58 | |
Powder |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
EINECSNumber : 205-814-1
RTECSNumber : VM2975000
TSCA : TSCA
RUO – Research Use Only