missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rubbermaid™ Xtra™ Black Utility Carts

Large shelves hold small baths, centrifuges, pH meters, and other equipment

Supplier:  Rubbermaid™ 409400GRAY

Catalog No. 11-926-42

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Black, four-shelf utility cart

  • Load capacity: 136.8kg (300 lb.)
  • Tough HDPE construction resists chips, dents, scratches, and rust
  • Easy-to-clean shelves with rounded interior corners and raised edges to keep small items from slipping off
  • Ergonomically designed handles on both cart ends
  • Quiet-rolling, nonmarking 10.2cm (4 in.) diia. casters

  • Choice of three-shelf models in three styles: open, enclosed on two ends, and enclosed on three sides (virtually a cabinet on wheels)
  • Also available in four-shelf open style

Order Info

Assembly required. No special tools needed. Three-shelf carts (L x W x H): 20 x 40.5 x 51 in. (51 x 103 x 130cm).

Specifications

Specifications

Instrument Cart
100 lb.
40.62 x 20 x 37.8 in. (103.17 x 50.8 x 96.01 cm)
37.8 in.
Lockable doors, enclosed end panels on 3 sides, 3 easy-to-clean shelves with rounded interior corners and raised edges, ergonomically handles, quiet-rolling nonmarking 4 in. dia. casters
103.17 cm
136 kg
3
50.8 cm
45.4 kg
Gray
Securely transporting tools and smaller equipment in hospitals, healthcare centers and labs
96.01 cm
40.62 in.
300 lb.
HDPE
20 in.
SDS
Documents

Documents

Promotions

Promotions