Thermo Scientific™ Rotor, 4-Place Swinging Bucket, for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ Centrifuge
For use with Legend Mach 1.6 and 1.6R Centrifuges
Manufacturer:  Thermo Scientific™ 75002000
Catalog No. 75002000
Description
Description
  • Rotor body only; buckets and biocontainment lids sold separately
  • Choose rectangular or round buckets
  • Adapters sold separately
Specifications
Specifications
Rotor
4,618 x g
Legend Mach 1.6 Series
4 x 400mL
4,700rpm
Documents