Learn More
Rifamycin SV sodium, 98+%, Thermo Scientific Chemicals
$173.90 - $634.10
Chemical Identifiers
CAS | 14897-39-3 |
---|---|
Molecular Formula | C37H47NNaO12 |
Molecular Weight (g/mol) | 720.77 |
MDL Number | MFCD09752825 |
InChI Key | YVOFSHPIJOYKSH-DITFEXNXSA-N |
PubChem CID | 124219641 |
IUPAC Name | sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1â´,â·.0âµ,²â¸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate |
SMILES | [Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6309503
|
Thermo Scientific Chemicals
J6309503 |
1 g |
Each for $173.90
|
|
|||||
AAJ6309506
|
Thermo Scientific Chemicals
J6309506 |
5 g |
Each for $634.10
|
|
|||||
Description
Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifamycin SV sodium is an antimicrobial agent used to inhibit the DNA polymerase and RNA polymerase in biochemistry. It is used for the treatment of HIV-related Tuberculosis, travelerâs diarrhea (TD), clostridium difficile-associated disease (CDAD) and rheumatoid knee synovitis. It is utilized to study potential genes like rif15 and rif16 during the conversion of rifamycin SV into rifamycin B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water, alcohol and dimethyl sulfoxide.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
14897-39-3 | |
720.77 | |
YVOFSHPIJOYKSH-DITFEXNXSA-N | |
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1â´,â·.0âµ,²â¸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate |
C37H47NNaO12 | |
MFCD09752825 | |
124219641 | |
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O |
Specifications
Rifamycin SV Sodium | |
14897-39-3 | |
MFCD09752825 | |
Soluble in water,alcohol and dimethyl sulfoxide. | |
[Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O | |
720.77 | |
719.45 | |
Orange to Red | |
Rifamycin SV sodium |
Powder | |
C37H47NNaO12 | |
14,8218 | |
YVOFSHPIJOYKSH-DITFEXNXSA-N | |
sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1â´,â·.0âµ,²â¸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate | |
124219641 | |
≥98% | |
1 g |
Safety and Handling
EINECSNumber : 238-965-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only