missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

pH Test Strips, Ricca Chemical

General-purpose pH test strips

$64.08 - $92.32

Specifications

For Use With (Application) For precise pH determination
Includes Color chart
Type pH Test Strip
View More Specs

Products 5
Catalog Number Mfr. No. pH Range Graduations Quantity Price Quantity  
Catalog Number Mfr. No. pH Range Graduations Quantity Price Quantity  
88-801
View Documents Promotion Details
Ricca Chemical Company
RI88801
0 to 14 1 1 Ea.
Each of 100 for $88.00
 
88-811
View Documents Promotion Details
Ricca Chemical Company
RI88811
0 to 6 0.5 100 Strips
Each for $92.32
 
88831
View Documents Promotion Details
Ricca Chemical Company
R8883000EACH
7 to 14 0.5 100 Strips
Each for $92.32
 
88-821
View Documents Promotion Details
Ricca Chemical Company
R8882000EACH
4.5 to 10 0.5 100 Strips
Each for $64.08
 
88-841
View Documents Promotion Details
Ricca Chemical Company
88841
2 to 9 0.5 1 Ea.
Each for $92.32
 
Description

Description

  • Available in four different pH ranges
  • Time-saving, single-use indicator strips with color-fixed test area on an easy-to-handle, durable plastic strip
  • Fast accurate results
  • Packages include a color comparison chart
  • 100 strips per pack

Specifications

Specifications

For precise pH determination
Color chart
pH Test Strip
SDS
missing translation for 'documents'

missing translation for 'documents'