Learn More
Thermo Scientific™ Restore™ Western Blot Stripping Buffer
Safely and effectively remove primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
Supplier: Thermo Scientific™ LSG21059
Description
Thermo Scientific™ Restore Western Blot Stripping Buffer safely and effectively removes primary and secondary antibodies from nitrocellulose and PVDF membranes to allow chemiluminescent Western blots to be reprobed.
Performing gel electrophoresis and duplicate immunoblot assays to test new primary antibodies or antibody concentrations is time-consuming and expensive. Restore Western Blot Stripping Buffer eliminates this waste when detecting immunoblots with chemiluminescent Western blotting substrates. The reagent allows clean and efficient removal of primary and secondary antibodies from immunoblots without removing or damaging the immobilized antigen. This allows blots to be stripped and reprobed with confidence.
Highlights:
Saves time – no need to re-run gels and blots
Saves costly sample – re-probe the membrane using the same target sample
Effective – formulation is more efficient at stripping antibodies than homemade buffers
Gentle – does not damage the target antigen during stripping allowing efficient reprobing
Odor-free – no mercaptans means no acrid odors
Economical – less expensive than other commercial stripping buffers
Reuse a nitrocellulose or PVDF blot to detect a different target with a different primary antibody; Reprobe a blot to correct or optimize antibody concentrations that were ineffective the first time.
Specifications
Western Blot Stripping Buffer | |
Liquid | |
Nitrocellulose, PVDF | |
500 mL |
Chemiluminescent | |
Western Blot | |
Restore™ | |
Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature. |
For Research Use Only. Not for use in diagnostic procedures.