Restek™ Resprep 12- or 24-Port Tube Manifold DFS Item
Use with any standard male luer end SPE cartridges.
Manufacturer:  Restek™ RK26077
Catalog No. 06713138
Includes: One each of glass basin with built-in vacuum regulator, polypropylene lid with 12 individual control valves, 12-position collection rack, PTFE sample guides, and waste container
Description
Description
  • Inert, PTFE sample guides reduce cross-contamination and carryover
  • Flexible sample collection rack will accommodate a variety of receiving vessels
  • Quick vacuum-release valve for better system control
  • Individual valves allow vacuum control for each cartridge, improving reproducibility
Specifications
Specifications
Standard male luer end SPE cartridges
Glass basin w/ built-in vacuum regulator, lid with 24 indiv. control valves, 24-pos. collection rack, PTFE sample guides
12
Resprep™ SPE manifold
Documents