Restek™ Replacement Split/Splitless Injection Ports: Replacement Weldments DFS Item
Replacement weldments for Agilent 5890 and 6890/6850 GCs
$1051.83 - $1800.94
Specifications
For Use With (Application) Gas Chromatography
View More Specs
Products 8
Catalog Number Mfr. No. For Use With (Equipment) Material Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Material Product Type Price Quantity  
06710552 DFS Item
View Documents Promotion Details
Restek™
RK20265
Manual Flow or Epc on Agilent 5890 GCs; Manual Flow Only on Agilent 6890/6850 GCs Stainless Steel Weldment
Each for $1,124.91
06710553 DFS Item
View Documents Promotion Details
Restek™
RK20266
Agilent 5890 GCs Stainless Steel Shell Weldment
Each for $1,192.47
06710554 DFS Item
View Documents Promotion Details
Restek™
RK20267
Manual Flow or Epc on Agilent 5890 GCs; Manual Flow Only on Agilent 6890/6850 GCs Siltek Treated Stainless Steel Weldment
Each for $1,352.36
06710555 DFS Item
View Documents Promotion Details
Restek™
RK20268
Agilent 5890 GCs Siltek Treated Stainless Steel Shell Weldment
Each for $1,363.47
06712018 DFS Item
View Documents Promotion Details
Restek™
RK22673
Agilent 6890/6850 GCs Stainless Steel Shell Weldmen
Each for $1,051.83
06712016 DFS Item
View Documents Promotion Details
Restek™
RK22671
Agilent 6890/6850 GCs Siltek Treated Stainless Steel Shell Weldment
Each for $1,263.55
06712017 DFS Item
View Documents Promotion Details
Restek™
RK22672
Agilent 6890/6850 GCs Siltek Treated Stainless Steel Weldment with EPC
Each for $1,800.94
06712019 DFS Item
View Documents Promotion Details
Restek™
RK22674
Agilent 6890/6850 GCs Stainless Steel Weldment with EPC
Each for $1,505.79
Description
Description
  • Siltek™ treatment passivates metal surface ensuring an inert pathway for the sample, delivering increased performance
  • High-quality stainless steel injector
  • Meets or exceeds Agilent original equipment performance
Specifications
Specifications
Gas Chromatography
SDS
missing translation for 'documents'
missing translation for 'documents'