Restek™ Opti-Cap DFS Item
Fits all standard GL-45 bottles.
Manufacturer:  Restek™ RK25300
Catalog No. 06712843
Description
Description
  • Economical way to helium-sparge and deliver LC mobile phases
  • Have two 0.125 in. holes and one 0.0625 in. hole for tubing
  • All three openings are designed for threaded PEEK plugs
Specifications
Specifications
Cap and PEEK plug
Opti-Cap™ Bottle
Documents