missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

VICI™ Restek™ Gas-Specific Purifiers DFS Item

Ideal for removing moisture, hydrocarbons and oxygen from gas chromatography gas supplies

$557.61 - $841.62

Specifications

Diameter (English) Outer 1.5 in.
Diameter (Metric) Outer 3.8cm
Product Type Gas purifier
Flow Rate 500mL/min.
For Use With (Application) Ideal for removing moisture, Hydrocarbons and Oxygen from gas chromatography gas supplies
View More Specs

Products 3
Catalog Number Mfr. No. Connector Types Gases Measured Price Quantity  
Catalog Number Mfr. No. Connector Types Gases Measured Price Quantity  
05-730-1 View Documents VICI™
MSP1001
0.12 in. Fitting Helium/inert Each for $841.62
05-730-5 View Documents VICI™
MSP3001
0.12 in. Fitting Nitrogen Each for $557.61
05-730-7 View Documents VICI™
P4001
0.12 in. Fitting Air Each for $814.97
Description

Description

Used in-line with the GC carrier gas or detector gas supply to produce gases that are at least a factor of ten higher in purity than a 99.9999% “Chromatography Grade” cylinder of gas (when the purifier is supplied by a 99.995% “High Purity” cylinder of gas).

  • Available with 1/8 in. or 1/4 in. compression fittings
  • All models feature proprietary adsorptive materials to help capture and retain specific contaminants for the operating life of the purifier
  • These materials adsorb a larger number and variety of contaminants than commonly used adsorptive materials

Specifications

Specifications

1.5 in.
Gas purifier
Ideal for removing moisture, Hydrocarbons and Oxygen from gas chromatography gas supplies
57.15cm
3.8cm
500mL/min.
22.5 in.
1000psi