Restek™ Flammable Gas Flash Arrestor - Factory Mutual Approved DFS Item
Valve cuts off gas supply in the event of a flashback.
Manufacturer:  Restek™ RK21334
Catalog No. 06711526
Description
Description
  • Can be reset and reused after flashback has occurred
  • Brass body
  • 0.25 in. Female NPT
  • 316 stainless steel valve spring
  • Polyethylene temperature sensor
  • Maximum pressure: Hydrogen, 50psig
  • Meets safety requirements of BS6158, EN730, ISO 5175, AS 4603 and are UL Listed 23Y5
Specifications
Specifications
BS6158, EN730, ISO 5175, AS 4603, UL Listed 23Y5
Flammable gases
Brass
Hydrogen Flashback Arrestor
Documents