Thermo Scientific™ Replacement Lid for Thermo Scientific™ 8 x 50mL Fixed Angle Rotor
Replacement Biocontainment Lid
Supplier: Thermo Scientific™ 75003011
Specifications
Swinging Bucket | |
Thermo Scientific Sorvall™ ST40/40R, SL40/40R, Sorvall Legend™ XT/XTR, ™ Megafuge 40, ™ Multifuge X3/X3R, Megafuge 40/40R Centrifuge Series | |
124 mm | |
Biocontainment certified by CAMR™, HPA, Porton Down, U.K. |