Thermo Scientific™ Internal Chamber Replacement Kit
For Use With: 310 Series (w/HEPA option) Incubators
Supplier: Thermo Scientific™ 190868
Description
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
- 99.97% filtering at 0.3μm
- Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
- Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
- Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
- Reduces downtime and labor spent on decontamination
Specifications
Internal chamber replacement kit with HEPA, gaskets, and other parts | |
310 Series (w/HEPA option) incubators | |
LED | |
Internal Chamber Replacement Kit |