missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Reichert™ Rhino Clinical Handheld VET360 Refractometer

Rugged portable refractometer for urine specific gravity and serum/plasma protein readings

Manufacturer:  Reichert™ 137536L0

Catalog No. 13-975-127


 

Includes: Manual has full set of conversion tables for protein, urine solids, and refractive index.

Description

Description

Rugged portable refractometer for urine specific gravity and serum/plasma protein readings

  • IP67 rated water resistant, dustproof, and impact resistant
  • Determines urine specific gravity and protein concentration in plasma, serum, and peritoneal fluid
  • Automatic temperature compensation provides accurate readings regardless of ambient temperature
  • Corrects all readings back to standard reference temperature of 20°C (68°F)—provides much greater accuracy than urinometers or reagent strips
  • Includes special scales for large and small animals
  • Completely sealed for indoor or outdoor use
  • Provides readings from 16° to 32°C (61° to 90°F)
To Use:
  • Place drop of sample on prism
  • Hold unit perpendicular to light source
  • Look into focusable eyepiece

Specifications

Specifications

±0.001 sp. gr., ±0.2g/100mL Serum Protein
Place drop of sample on prism, hold unit perpendicular to light source, look into focusable eyepiece
IP67
Manual has full set of conversion tables for Protein, Urine solids and refractive index
9.2 in.
1.6 in.
9.2 x 1.6 x 1.6 in. (233 x 40.6 x 40.6mm)
1 to 1.06 USG, 2 to 14g/100mL Serum Protein
1.6 in.
40.6mm
0.001 sp. gr., 0.1g/100mL Serum Protein
233mm
40.6mm
Documents