MilliporeSigma™ Reagents for Amicon™ Pro Affinity Concentration Kit Protein A with Amicon™ Ultra-0.5 Device
Reagents for MilliporeSigma™ Amicon Pro Affinity Concentration Kit Protein A with Amicon Ultra–0.5 Device
Supplier: MilliporeSigma™ ACR5000PA
Description
For use with EMD Millipore Amicon Pro Affinity Concentration Kit Protein A with Amicon Ultra–0.5 Device (Cat. No. ACK5003PA)
- 3mL Protein A resin (Binding capacity: 20mg human IgG/mL agarose)
- 2 X 66mL Bind/Wash Buffer
- 12mL Elution buffer
- 2.5mL Neutralization buffer
Specifications
Amicon Pro Affinity | |
3mL Protein A resin, 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer | |
Protein A Reagent Kit |
Antibody purification | |
1/Pk. |