Learn More
(R)-(-)-2-Octanol, 99%, Thermo Scientific Chemicals
$93.51 - $276.16
Chemical Identifiers
CAS | 5978-70-1 |
---|---|
Molecular Formula | C8H18O |
Molecular Weight (g/mol) | 130.231 |
MDL Number | MFCD00064284 |
InChI Key | SJWFXCIHNDVPSH-MRVPVSSYSA-N |
Synonym | r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol |
PubChem CID | 80080 |
ChEBI | CHEBI:37871 |
IUPAC Name | (2R)-octan-2-ol |
SMILES | CCCCCCC(C)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1150203
|
Thermo Scientific Chemicals
L1150203 |
1 g |
Each for $93.51
|
|
|||||
AAL1150206
|
Thermo Scientific Chemicals
L1150206 |
5 g |
Each for $276.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5978-70-1 | |
130.231 | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
80080 | |
(2R)-octan-2-ol |
C8H18O | |
MFCD00064284 | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
CHEBI:37871 | |
CCCCCCC(C)O |
Specifications
5978-70-1 | |
179°C to 180°C | |
Pleasant | |
1.426 | |
1 g | |
14,6752 | |
SJWFXCIHNDVPSH-MRVPVSSYSA-N | |
CCCCCCC(C)O | |
130.231 | |
CHEBI:37871 | |
99% |
0.819 | |
76°C (169°F) | |
C8H18O | |
MFCD00064284 | |
1719324 | |
r-octan-2-ol, r---2-octanol, r-2-octanol, l-octan-2-ol, 2r-octan-2-ol, 2r-2-octanol, unii-2shb67xf6h, d-2-octanol, r---hexylmethylcarbinol, l--2-octanol | |
−10° (c=1 in Ethanol) | |
(2R)-octan-2-ol | |
80080 | |
130.23 | |
(R)-(-)-2-Octanol |
Safety and Handling
GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: III
EINECSNumber : 227-777-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only