missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Applied Biosystems™ QuantStudio™ 5 Real-Time PCR System (0.1mL), 96-well and TaqMan Master Mix DFS Item

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems(TM) QuantStudio Real-Time PCR Systems, combined with Applied Biosystems(TM) TaqMan Master Mix bundle offers exceptional specificity with the speed and ease-of-use of Cloud connectivity.

Manufacturer:  Applied Biosystems™ LSA38707

Catalog No. A38707

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

The Applied Biosystems™ QuantStudio™ 3 and 5 Real-Time PCR Systems are the latest additions to our family of high-performance QuantStudio instruments. They provide a modern usability experience similar to mobile devices, allowing you to connect to your data easier than ever before. Therefore, you can access, analyze, and share data anytime, anywhere; and support remote monitoring of your runs, quick analysis of sophisticated data sets, secure storage of your results, and safe sharing of results online with colleagues across campus and around the world with web browser-based software supported by the Thermo Fisher Cloud platform.

For researchers performing gene expression who need an intuitive and easy to use qPCR solution, the Applied Biosystems QuantStudio Real-Time PCR Systems, combined with Applied Biosystems™ TaqMan® Master Mix bundle offers exceptional specificity with the speed and ease-of-use of Cloud connectivity.

This bundle includes:

  • QuantStudio 5 qPCR System, 96-well, 0.1 mL, Laptop
  • SuperScript IV VILO Master Mix with ezDNase (50 rxn)
  • TaqMan® Fast Advanced Master Mix (5 x 5 mL)
  • MicroAmp Fast Optical 96-Well Reaction Plate, 0.1 mL (10 plates)
  • MicroAmp Optical Adhesive Film (100 covers)
Specifications

Specifications

27cm
40cm
20 in.
26kg
100-240 V
Non-Interchangable
0.4°C
4 to 100.0°C
1 Year
QuantStudio Real Time PCR System (qPCR)
±3.66°C/sec
96-well, 0.1-mL block
50cm
10.5 in.
16 in.
57 lb
960 W
9.0°C/sec
0.25°C
Touchscreen
qPCR
50cm
4 to 100.0°C
No
Documents
Promotions

Promotions

  • Choose Applied Biosystems™ QuantStudio™ Real-Time PCR Systems for an easy-to-use qPCR solution. Bundle your system with an Applied Biosystems™ SYBR™ Green Master Mix or TaqMan® Master Mix to get both exceptional specificity and the speed of cloud connectivity.

    12/31/19

  • Choose Applied Biosystems QuantStudio Real-Time PCR Systems for an easy-to-use qPCR solution. Bundle your system with an Applied Biosystems SYBR Green Master Mix or TaqMan Master Mix to get both exceptional specificity and the speed of cloud connectivity.

    12/31/19