Learn More
Pyridine, Ultrapure, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
$88.45 - $916.95
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA32436AK
|
Thermo Scientific Chemicals
032436AK |
250 mL |
Each for $88.45
|
|
|||||
AA32436K2
|
Thermo Scientific Chemicals
032436K2 |
1 L |
Each for $255.67
|
|
|||||
AA32436M1
|
Thermo Scientific Chemicals
032436M1 |
4 x 1 L |
Each for $916.95
|
|
|||||
AA32436K7
|
Thermo Scientific Chemicals
032436K7 |
4 L |
Each for $681.77
|
|
|||||
Description
Pyridine is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
°C to 42°C | |
Colorless | |
0.978 | |
250 mL | |
Unpleasant | |
1.51 | |
C5H5N | |
20 hPa (15mm Hg) at 20°C | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 | |
Pyridine |
110-86-1 | |
Colorless liquid | |
115°C | |
20°C (68°F) | |
Liquid | |
>99.5% | |
MFCD00011732 | |
103233 | |
Miscible with water,alcohol,ether,pet ether,and oils. | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
Spectrophotometric |
Safety and Handling
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302+H312+H332-H315-H319
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 203-809-9
RTECSNumber : UR8400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only