Learn More
Pyridine (Certified ACS), Fisher Chemical™
$382.18 - $1725.09
Chemical Identifiers
CAS | 110-86-1 |
---|---|
Molecular Formula | C5H5N |
Molecular Weight (g/mol) | 79.102 |
MDL Number | MFCD00011732 |
InChI Key | JUJWROOIHBZHMG-UHFFFAOYSA-N |
Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
PubChem CID | 1049 |
ChEBI | CHEBI:16227 |
IUPAC Name | pyridine |
SMILES | C1=CC=NC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
P368-500
|
Thermo Fisher Scientific
P368500 |
500 mL | Glass Bottle |
Each for $382.18
Case of 6 Each for $2,293.08
|
|
||||
P368-4
|
Thermo Fisher Scientific
P3684 |
4 L | Glass Bottle |
Each for $1,725.09
Case of 4 Each for $6,900.36
|
|
||||
Chemical Identifiers
110-86-1 | |
79.102 | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
1049 | |
pyridine |
C5H5N | |
MFCD00011732 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
CHEBI:16227 | |
C1=CC=NC=C1 |
Specifications
8.5 | |
110-86-1 | |
0.978g/cm³ | |
500 mL | |
Glass Bottle | |
0.002% max. | |
C5H5N | |
MFCD00011732 | |
0.95 mPa/s at 20°C | |
0.1% max. | |
JUJWROOIHBZHMG-UHFFFAOYSA-N | |
pyridine | |
1049 | |
79.1 | |
Suitable for Karl Fischer reagent |
-42°C | |
Colorless | |
115°C | |
Pass Test | |
Liquid | |
≥99 % | |
N:CHCH:CHCH:CH | |
20mbar at 20°C | |
1282 | |
azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 | |
C1=CC=NC=C1 | |
79.102 | |
CHEBI:16227 | |
≥99% | |
Pyridine |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Harmful by inhalation, in contact with skin and if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:2
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid