Corning™ PYREX™ Screw Cap Culture Tubes with PTFE Lined Phenolic Caps
PTFE interface is firmly bonded to glued-in rubber liner
$530.69 - $1822.78
Specifications
Closure Color Black
Closure Material Phenolic
Design Type Threaded
Material Borosilicate Glass
Label Type Marking Spot
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Length (Metric) Thread Size Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Length (Metric) Thread Size Price Quantity  
14-933-1B
View Documents Promotion Details
Corning™
982620X
34 mL 20 mm 150 mm 18-415
Case of 192 Each for $1,822.78
14-932H
View Documents Promotion Details
Corning™
982616
11 mL 16 mm 100 mm 15-415
Pack of 144 Each for $975.45
Case of 2 Pack for $1,950.90
14-933-1A
View Documents Promotion Details
Corning™
982616X
15 mL 16 mm 125 mm 15-415
Pack of 144 Each for $963.91
Case of 2 Pack for $1,927.82
14-933-1C
View Documents Promotion Details
Corning™
982625X
70 mL 25 mm 200 mm 24-410
Pack of 48 Each for $601.03
Case of 3 Pack for $1,803.09
14-933A
View Documents Promotion Details
Corning™
982613
9 mL 13 mm 100 mm 13-415
Pack of 144 Each for $816.83
Case of 2 Pack for $1,633.66
14-933C
View Documents Promotion Details
Corning™
982620
25 mL 20 mm 125 mm 18-415
Pack of 96 Each for $866.69
Case of 2 Pack for $1,733.38
14-933D
View Documents Promotion Details
Corning™
982625
50 to 55 mL 25 mm 150 mm 24-410
Pack of 48 Each for $530.69
Case of 3 Pack for $1,592.07
Description
Description
Similar to Cat. No. 9825, but phenolic cap has a PTFE interface firmly bonded to the glued-in rubber liner, providing a sure and highly inert sealing face.
For phenolic caps, see Cat. No. 9998.
Specifications
Specifications
Black
Threaded
Marking Spot
Non-sterile
Screw Cap
Phenolic
Borosilicate Glass
PTFE
No
Culture Tube
SDS
missing translation for 'documents'
missing translation for 'documents'