missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

PYREX™ 24/29 Standard Taper Stopcock Vacuum-Rated Glass Desiccators, Complete (Bowl and Cover)

Desiccators have a rugged 24/29 standard taper stopcock with a tooled hose connection that serves as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum.

$705.06 - $1,460.74

Specifications

Diameter (English) Outside Tube 0.39 in.
Diameter (Metric) Outside Tube 1cm
Material Borosilicate Glass
Product Type Desiccator
Quantity Each
View More Specs

Alert:

Do not heat or subject to pressure. Rated to 1-atmosphere vacuum.


Includes: Cover, bowl, and stopcock.

Products 3
Catalog Number Mfr. No. Capacity (English) Depth (English) Height (English) Size Price Quantity  
Catalog Number Mfr. No. Capacity (English) Depth (English) Height (English) Size Price Quantity  
08-624-420 Accessories View Documents Corning Incorporated
C3121150
0.63 gal. 3.18 in. 9.4 in. Small Case for $705.06
08-624-424 Accessories View Documents Corning Incorporated
C3121250
2.77 gal. 4.72 in. 13.5 in. Large Case for $1,408.94
08-624-422 Accessories View Documents Corning Incorporated
C3121200
1.53 gal. 4.5 in. 11.6 in. Large Case for $1,460.74
Description

Description

Desiccators have a rugged 24/29 standard taper stopcock with a tooled hose connection that serves as the vacuum inlet. It is designed so that its manipulation is not affected by the vacuum.

  • Tubulation O.D. approx. 12mm
  • Borosilicate glass 7740
  • Design eliminates need for grease

Order Info

Replacement bowls and covers available separately. See Corning PYREX Brand Desiccator Replacement Covers and Corning PYREX Brand Desiccator Replacement Bowls. For replacement standard taper stopcock, see Cat. No. 08-624-426.

Specifications

Specifications

0.39 in.
Borosilicate Glass
Each
PTFE
1cm
Desiccator
Cover, bowl and stopcock, 24/29 standard taper external sleeves with tooled hose connections