missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Propane, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S3145

Catalog No. S-3145



Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Propane74-98-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Propane
2mL amber prescored glass ampule
0.1,99.9
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
C3H8
CCC
44.097
CHEBI:32879
74-98-6,67-56-1
1 mL
0.1,99.9
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
ATUOYWHBWRKTHZ-UHFFFAOYSA-N
propane
6334
Methanol-P&T

Chemical Identifiers

74-98-6
44.097
6334
propane
C3H8
ATUOYWHBWRKTHZ-UHFFFAOYSA-N
CHEBI:32879
CCC
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only