missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Steady-Glo™ Luciferase Assay System
Steady-Glo™ Luciferase Assay System is a homogeneous assay with signal half-life of over 5 hours in commonly used cell culture media. Ideal for batch processing.
$214.53 - $1,542.52
Specifications
Detection Method | Luminescence |
---|---|
For Use With (Equipment) | 96- and 384-well plates |
Content And Storage | -30°C to -10°C |
For Use With (Application) | For quantitation of firefly luciferase expression in mammalian cells |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR-E2520
|
Promega
E2520 |
100 mL |
N/A
|
|
|||||
PR-E2510
|
Promega
E2510 |
10 mL |
N/A
|
|
|||||
PR-E2550
|
Promega
E2550 |
10 x 100 mL |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Long-Lived Luminescence for Batch Processing in Multiwell Plates
- Stable luminescence signal with 5-hour half-life
- No need to remove culture medium or wash cells prior to adding assay reagent
- Less sensitive to mixing conditions in multiwell plates. Particularly useful in 384-well plates
This product does not convey a license to use recombinant Renilla luciferase under U.S. Pat. No. 5,418,155 and related patents. Promega sells licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Certain applications of this product may require licenses from others.
Patent Pending.
Specifications
Luminescence | |
96- and 384-well plates | |
-30°C to -10°C | |
For quantitation of firefly luciferase expression in mammalian cells |