missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Promega™ Kinase-Glo™ Luminescent Kinase Assay DFS Item

Homogeneous, nonradioactive method of determining the activity of kinases

$161.58 - $4,560.33

Specifications

Detection Method Homogeneous, Nonradioactive Method
Product Type Kinase-Glo Luminescent Kinase Assay
For Use With (Application) For determining the activity of purified Kinases by quantifying the amount of ATP
For Use With (Equipment) For multiwell plates
Storage Requirements -20°C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PRV6714 View Documents Promega™
V6714
10 x 100mL Pack of 10 Each for $4,560.33
PR-V6711 View Documents Promega™
V6711
10mL Each for $161.58
PR-V6712 View Documents Promega™
V6712
10 x 10mL Each for $724.38
PR-V6713 View Documents Promega™
V6713
100mL Each for $612.03
Description

Description

  • Quantifies the amount of ATP remaining in solution following a kinase reaction1, 2
  • Designed for use with multiwell plate formats, making it ideal for high throughput screening
  • Can be used with a wide range of kinases, including lipid kinases and substrates
  • Features low susceptibility to library compounds; Z' values are >0.8 in either 96- or 384-well formats

1U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
2The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024, 5,674,713 and 5,700,673.

Specifications

Specifications

Homogeneous, Nonradioactive Method
For determining the activity of purified Kinases by quantifying the amount of ATP
-20°C
Kinase-Glo Luminescent Kinase Assay
For multiwell plates