Promega dNTP Mix
A premixed solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10mM in water.
Supplier: Promega U1511
Description
- Ready to Use Solution of Deoxynucleotide Triphosphates
- Highly pure (>99% triphosphate)
- 10mM each nucleotide in water
- Reduce pipetting steps by adding nucleotides as a mix
Specifications
7.5 | |
10 mM | |
200 μL |
-30°C to -10°C | |
>99% | |
PCR amplification |