missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega CytoTox-ONE™ Homogeneous Membrane Integrity Assay
A fast homogeneous, fluorometric method for estimating the number of non-viable cells present in multiwell plates. Based on detection of LDH release, an indicator of cytotoxicity.
$298.76 - $933.13
Specifications
Detection Method | Fluorometric |
---|---|
For Use With (Application) | Cytotoxicity determination, total cell number determination, measures release of Lactate Dehydrogenase |
Format | 96- or 384-well Plates |
Content And Storage | -30°C to -10°C |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity | ||||
PR-G7890
|
Promega
G7890 |
CytoTox-ONE™ Homogeneous Membrane Integrity Assay | 200 to 800 Assays |
Each for $298.76
|
|
||||
PR-G7891
|
Promega
G7891 |
CytoTox-ONE™ Homogeneous Membrane Integrity Assay | 1000 to 4000 Assays |
Each for $933.13
|
|
||||
Description
- A Fluorometric Method for Estimating the Number of Nonviable Cells Present in Multiwell Plates
- Elimination of cell transfer step saves time
- Perform multiple assays on one sample
Specifications
Fluorometric | |
Cytotoxicity determination, total cell number determination, measures release of Lactate Dehydrogenase | |
96- or 384-well Plates | |
-30°C to -10°C |
U.S. Pat. Nos. 6,982,152 and 7,282,348, Australian Pat. No. 2003223521 and other patents pending.