missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega cAMP-Glo™ Max Cyclic-AMP Assay
The cAMP-Glo™ Max Assay is a homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells. Compounds that modulate GPCRs coupled with adenylate cyclase typically alter intracellular cAMP levels.
$718.60 - $3961.57
Specifications
Content And Storage | -30°C to -10°C |
---|---|
For Use With (Application) | A homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells |
Type | Detection Assay Kit |
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
PRV1681
|
Promega
V1681 |
100μL cAMP (1mM), 1mL MgCl2 (1M), 2.5mL cAMP-Glo™ ONE-buffer, 20μL protein Kinase A, 1 bottle Kinase-Glo substrate (lyophilized), 10mL Kinase-Glo buffer | 2 Preps |
Each for $718.60
|
|
||||
PRV1682
|
Promega
V1682 |
500μL cAMP (1mM), 5mL MgCl2 (1M), 20mL cAMP-Glo™ ONE-buffer, 200μL protein Kinase A, 1 bottle Kinase-Glo substrate (lyophilized), 100mL Kinase-Glo buffer | 20 Preps |
Each for $3,961.57
|
|
||||
Description
- Faster cAMP Detection for High-Throughput Assays
- Complete in ∼30 minutes
- Scalable to 1536-well plate formats and beyond
- No interference by fluorescent compounds
Specifications
-30°C to -10°C | |
A homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells | |
Detection Assay Kit |