missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Promega Beta-Glo™ Assay System
The Beta-Glo™ Assay System is a homogeneous, luminescence-based method for quantitating beta-galactosidase expression in mammalian cells.
$232.95 - $10480.40
Specifications
pH | 7.5 |
---|---|
Content And Storage | -30°C to -10°C |
Form | Liquid |
Includes: Beta-Glo Assay Buffer; Beta-Glo Assay Substrate
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR-E4720
|
Promega
E4720 |
10 mL |
Each for $232.95
|
|
|||||
PRE4740
|
Promega
E4740 |
100 mL |
Each for $1,663.60
|
|
|||||
PRE4780
|
Promega
E4780 |
10 x 100 mL |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Luminescence Detection of β-Gal Activity
- Add a single reagent directly to cells in growth medium
- Optimal assay performance at room temperature
- Read results using any luminometer. Injectors not required
Specifications
7.5 | |
-30°C to -10°C | |
Liquid |