missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Productainer™ BPC Bag with 3 Ports

Using these BPC bags with 3 ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

$214.51 - $244.03

Specifications

Description Two-dimensional disposable cell culture/bioprocess bag; Size: 200L; Fittings: MPC insert, MPC Body, Female Luer Lock; with tubing
Film Description CX3-9 Film
Port Detail 3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in.Female Luer Lock
Storage Requirements Ambient
Washdown Line No
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
SH3066702 View Documents Thermo Scientific™
SH3066702
100L Each for $233.97
SH3066703 View Documents Thermo Scientific™
SH3066703
200L Each for $244.03
SH3066701 View Documents Thermo Scientific™
SH3066701
50L Each for $214.51
Description

Description

Using Thermo Scientific™ Productainer™ BPC bag with 3 ports, package, store, and transport 50 to 200L volumes of media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific CX5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Highlights:

  • Three sizes - 50, 100 and 200L
  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components

Recommended for:

  • Harvest - Collection and storage of bulk product from bioreactors

  • Storage - Sterile storage of media, buffers and process intermediates

  • Separation - Feeding and receiving of sterile liquids from separation systems

  • Purification - Buffer feeding to purification system, fraction collection and storage

  • Filling - Sterile reservoir for filling systems

  • Transport and Shipping - Bulk sterile liquids, buffers, media and process intermediates within and between facilities

  • Pillow Design, 2-dimensional
  • Constructed with Thermo Scientific CX5-14 film with a contact surface composed of ultra-low density polyethylene
  • Three face ports (3 to 9.5mm (3/8in.))
  • Line 1: 9.5mm (3/8in.) MPC insert constructed of polycarbonate with 61cm (24in.) of C-Flex™ SEBS tubing (9.5mm (3/8in.) ID by 15.9mm (5/8in.) OD)
  • Line 2: 9.5mm (3/8in.) MPC Body constructed of polycarbonate with 61cm (24in.) of C-Flex SEBS tubing (9.5mm (3/8in.) ID by 15.9mm (5/8in.) OD)
  • Line 3: 6.4mm (1/4in.) Female Luer Lock constructed of polypropylene with 66cm (26in.) of C-Flex SEBS tubing (6.4mm (1/4in.) ID by 9.5mm (3/8in.) OD)
  • Designed to be used with flat bottom linear low density polyethylene drums with clamps and matching lids
  • Plastic drum dolly for ergonomic convenience
Specifications

Specifications

Two-dimensional disposable cell culture/bioprocess bag; Size: 200L; Fittings: MPC insert, MPC Body, Female Luer Lock; with tubing
3/8in. Quick Disconnect Insert; 3/8in. Quick Disconnect Body; 1/8in.Female Luer Lock
No
CX3-9 Film
Ambient