missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polypropylene Disposable Beakers
Disposable or steam-autoclavable for reuse.
$45.90 - $127.41
Specifications
Material | Polypropylene |
---|---|
Includes | Spout |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Single-use |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S63359
|
Customer Channels Group (Ccg)
L1210EMD |
30 mL |
Pack of 100 Each for $45.90
Case of 10 Pack for $459.00
|
|
|||||
S63360
|
Customer Channels Group (Ccg)
L1220EMD |
50 mL |
Pack of 100 Each for $51.56
Case of 10 Pack for $515.60
|
|
|||||
S63361
|
Customer Channels Group (Ccg)
L1240EMD |
150 mL |
Pack of 100 Each for $127.41
Case of 10 Pack for $1,274.10
|
|
|||||
S63362
|
Customer Channels Group (Ccg)
L1260EMD |
400 mL |
Pack of 50 Each for $109.38
Case of 10 Pack for $1,093.80
|
|
|||||
Description
- Resistant to most acids, solvents and other common reagents
- Special “dripless” lip
- Rib construction permits stacking without binding
- Graduated markings in mL and oz.
Specifications
Polypropylene | |
Beaker | |
Single-use |
Spout | |
Autoclavable | |
Yes |