missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Justrite™ Polyethylene Corrosives/Acid Cabinet

Sturdy, completely metal-free poly cabinet for storing acids or bases without the worry of corrosion

Manufacturer:  Justrite™ JMC24180

Catalog No. 19-414-063


Alert:

Research environments using many different types of chemicals have special storage requirements. Whether using acids, bases or solvents, it is critical that incompatible liquids are segregated to avoid disastrous explosions or fire. The SDS (Safety Data Sheet) should be reviewed to determine chemical characteristics and recommended storage practices


Description

Description

Safely store strong acids such as hydrochloric, sulfuric or nitric acid; or bases such as sodium, potassium or calcium hydroxide in this durable, HDPE (high-density polyethylene) cabinet. With no metal parts to corrode, this cabinet holds up to 36, 2.5L (90L) bottles, and offers chemical resistance against spills or potential damaging vapors when storing acids or bases. The shelf adjusts on 3 in. (76-mm) increments, and has a load capacity of 125 lb. (57kg); choose from three positions to optimize container storage. A removable yet sturdy sump cover, which also serves as a bottom shelf, conceals the fully integrated, leakproof sump with an 8.5-gal (32L) capacity. Recessed channels in shelf and base help direct spills to the back of the cabinet and down into the sump. Three application-specific labels identify contents to safely store and segregate incompatible chemicals — ACID, BASE, and CORROSIVE. Doors open nearly 180 degrees for easy access, engage firmly with the body when closed, and accept a padlock (not included) for added security.
Specifications

Specifications

36, 2.5L (90L) bottles
457mm
Store acids or bases
889mm
2
914mm
Blue
HDPE (High-density Polyethylene)
18 in.
Manual-close
28 in.
711mm
31.25 in.
794mm
35 in.
36 in.
Documents