GSC Go Science Crazy Pinchcock Attachment for Buret
Tubing slips over the end of any buret.
Supplier: GSC Go Science Crazy EPD2051
Description
Attachment includes a glass delivery tip, pinchcock and short piece of rubber tubing.
Specifications
Glass Delivery Tip, Pinchcock, Rubber Tube | |
Pinchcock Attachment |